• USA Home
  • Search Results

  • orthophosphoricacid859800766438211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

ortho-Phosphoric acid 85%

4 Products
Synonym:   Orthophosphoric acid, Phosphoric acid, o-Phosphoric acid, Orthophosphoric acid
Image of ortho-Phosphoric acid 85%
CAS Number:
Linear Formula:
 H3PO4
Molecular Weight:
 98.00
 Beilstein/REAXYS Number:
 1921286
 E Number:
 E338
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Viscosity
  Agency/Method
  Special Grade
  pH Value
Show More