• USA Home
  • Search Results

  • osmiumtetroxidesolution254232081612011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Osmium tetroxide solution

4 Products
Image of Osmium tetroxide solution
CAS Number:
Linear Formula:
 OsO4
Molecular Weight:
 254.23
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Special Grade
  Color
  Density
  Vapor pressure
  Application(s)
Show More