• USA Home
  • Search Results

  • otoluenesulfonamide171228819711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

o-Toluenesulfonamide

3 Products
Image of <I>o</I>-Toluenesulfonamide
CAS Number:
Linear Formula:
 CH3C6H4SO2NH2
Molecular Weight:
 171.22
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
Show More