• USA Home
  • Search Results

  • otoluidine107159553411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

o-Toluidine

5 Products
Synonym:   2-Aminotoluene, 2-Methylaniline
Image of <I>o</I>-Toluidine
CAS Number:
Linear Formula:
 CH3C6H4NH2
Molecular Weight:
 107.15
 Beilstein/REAXYS Number:
 741981
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Special Grade
  pH Value
  Vapor pressure
  Transition temperature
  Potency
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Application(s)
  Physical Form
  Assay
Show More