• USA Home
  • Search Results

  • oxalicacid900314462711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oxalic acid

6 Products
Image of Oxalic acid
CAS Number:
Linear Formula:
 HO2CCO2H
Molecular Weight:
 90.03
 Beilstein/REAXYS Number:
 385686
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
  Purity Grade
  Melting Point
  Special Grade
Show More