• USA Home
  • Search Results

  • oxaliplatin397296182594311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oxaliplatin

4 Products
Synonym:   [SP-4-2-(1R-trans)]-(1,2-Cyclohexanediamine-N,N′)[ethanedioata(2--)-O,O’]platinum
Image of Oxaliplatin
CAS Number:
Empirical Formula (Hill Notation):
 C8H14N2O4Pt
Molecular Weight:
 397.29
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Manufacturer designation
Show More