• USA Home
  • Search Results

  • oxalylchloride126937937811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oxalyl chloride

3 Products
Synonym:   Oxalic acid dichloride, Oxalic acid dichloride
Image of Oxalyl chloride
CAS Number:
Empirical Formula (Hill Notation):
 C2Cl2O2
Molecular Weight:
 126.93
 EC Index Number
 201-200-2
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More