• USA Home
  • Search Results

  • oxotremorinesesquifumaratesalt38039anhydrousbasis1736035911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oxotremorine sesquifumarate salt

1 Products
Synonym:   1-(4-[1-Pyrrolidinyl]-2-butynyl)-2-pyrrolidinone, 1-(4-[1-Pyrrolidinyl]2-butynyl)-2-pyrrolidinone
Image of Oxotremorine sesquifumarate salt
CAS Number:
Empirical Formula (Hill Notation):
 C12H18N2O · 1.5C4H4O4 · xH2O
Molecular Weight:
 380.39 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More