• USA Home
  • Search Results

  • oxybutyninchloride39395150865211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oxybutynin chloride

4 Products
Synonym:   α-Phenylcyclohexaneglycolic acid 4-(diethylamino)-2-butynyl ester hydrochloride
Image of Oxybutynin chloride
CAS Number:
Empirical Formula (Hill Notation):
 C22H31NO3 · HCl
Molecular Weight:
 393.95
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More