• USA Home
  • Search Results

  • oxylene106179547611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

o-Xylene

9 Products
Synonym:   1,2-Dimethylbenzene
Image of o-Xylene
CAS Number:
Linear Formula:
 C6H4(CH3)2
Molecular Weight:
 106.17
 EC Index Number
 202-422-2
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Refractive Index
  Anal. Standard Form
  Physical Form
  UV-Absorption Condition
  Titertest
  Solubility
  Assay
  UV-Absorption Values
Show More