• USA Home
  • Search Results

  • oxypeucedanin2862873752011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oxypeucedanin

1 Products
Synonym:   4-[[(2S)-3,3-Dimethyl-2-oxiranyl]methoxy]-7H-furo[3,2-g][1]benzopyran-7-one
Image of Oxypeucedanin
CAS Number:
Empirical Formula (Hill Notation):
 C16H14O5
Molecular Weight:
 286.28
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More