• USA Home
  • Search Results

  • oxypeucedaninhydrate30429264385811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oxypeucedanin hydrate

1 Products
Synonym:   Aviprin, Hydroxypeucedanin hydrate, Prangol, Prangolarin hydrate
Image of Oxypeucedanin hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C16H16O6
Molecular Weight:
 304.29
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More