• USA Home
  • Search Results

  • p1p5diadenosine5pentaphosphatetrilithiumsalt934177552297311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

P1,P5-Di(adenosine-5′) pentaphosphate trilithium salt

1 Products
Synonym:   A(5′)P5(5′)A trilithium salt, Diadenosine pentaphosphate trilithium salt
Image of P<SUP>1</SUP>,P<SUP>5</SUP>-Di(adenosine-5′) pentaphosphate trilithium salt
CAS Number:
Empirical Formula (Hill Notation):
 C20H26Li3N10O22P5
Molecular Weight:
 934.17
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 32160414
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More