• USA Home
  • Search Results

  • paclitaxel853913306962411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Paclitaxel

6 Products
Image of Paclitaxel
CAS Number:
Empirical Formula (Hill Notation):
 C47H51NO14
Molecular Weight:
 853.91
 Beilstein/REAXYS Number:
 1420457
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
  Manufacturer designation
  Color
  Original Developer
  Physical Form
  Biological Source
  Assay
  Mode of Action
  Activity Spectrum
  Special Grade
Show More