• USA Home
  • Search Results

  • paliperidone4264814459875411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Paliperidone

2 Products
Synonym:   3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 9-Hydroxyrisperidone
Image of Paliperidone
CAS Number:
Empirical Formula (Hill Notation):
 C23H27FN4O3
Molecular Weight:
 426.48
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More