• USA Home
  • Search Results

  • palladiumiiacetatechemdosetablets22451337531311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Palladium(II) acetate, ChemDose tablets

1 Products
Synonym:   ChemDose, Palladium(II) acetate tablets, Palladium(II) acetate impregnated tablets, Pd(OAc)2 impregnated tablets
Image of Palladium(II) acetate, ChemDose<SUP>™</SUP> tablets
CAS Number:
Linear Formula:
 Pd(OCOCH3)2
Molecular Weight:
 224.51
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
Show More