• USA Home
  • Search Results

  • palladiumiitrifluoroacetate332454219631611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Palladium(II) trifluoroacetate

1 Products
Synonym:   Pd(TFA)2, Trifluoroacetic acid palladium(II) salt
Image of Palladium(II) trifluoroacetate
CAS Number:
Linear Formula:
 (CF3COO)2Pd
Molecular Weight:
 332.45
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More