• USA Home
  • Search Results

  • palmiticacid113c257425767753911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Palmitic acid-1-13C

2 Products
Synonym:   Hexadecanoic acid-1-13C
Image of Palmitic acid-1-<SUP>13</SUP>C
CAS Number:
Linear Formula:
 CH3(CH2)1413CO2H
Molecular Weight:
 257.42
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
Show More