• USA Home
  • Search Results

  • palmitoleicacid2544137349911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Palmitoleic acid

2 Products
Synonym:   cis-9-Hexadecenoic acid, cis-9-Palmitoleic acid
Image of Palmitoleic acid
CAS Number:
Linear Formula:
 CH3(CH2)5CH=CH(CH2)7COOH
Molecular Weight:
 254.41
 Beilstein/REAXYS Number:
 1725389
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Titertest
Show More