• USA Home
  • Search Results

  • pancreatinfromporcinepancreas12345804947611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pancreatin from porcine pancreas

4 Products
Synonym:   Pancreatin from hog pancreas
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.54
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Agency/Method
  Additive
  Specific Activity
Show More