• USA Home
  • Search Results

  • pancuroniumbromide732671550066011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pancuronium bromide

4 Products
Synonym:   1,1′-[(2β,3α,5α,16β,17β)-3,17-bis(Acetyloxy)androstane-2,16-diyl]bis(1-methylpiperidinium) dibromide
Image of Pancuronium bromide
CAS Number:
Empirical Formula (Hill Notation):
 C35H60Br2N2O4
Molecular Weight:
 732.67
 Beilstein/REAXYS Number:
 4226892
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More