• USA Home
  • Search Results

  • panisaldehyde1361512311511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

p-Anisaldehyde

6 Products
Synonym:   4-Methoxybenzaldehyde, p-Methoxybenzaldehyde, Aubépine
Image of <I>p</I>-Anisaldehyde
CAS Number:
Linear Formula:
 CH3OC6H4CHO
Molecular Weight:
 136.15
 Beilstein/REAXYS Number:
 471382
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Anal. Standard Form
  Allergen
  Organoleptic property
  Shelf Life
  Assay
  Titertest
  Physical Form
  Special Grade
Show More