• USA Home
  • Search Results

  • papaverinehydrochloride375856125611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Papaverine hydrochloride

7 Products
Synonym:   6,7-Dimethoxy-1-veratrylisoquinoline hydrochloride
Image of Papaverine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C20H21NO4 · HCl
Molecular Weight:
 375.85
 Beilstein/REAXYS Number:
 3921435
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
  Anal. Standard Form
  Physical Form
Show More