• USA Home
  • Search Results

  • paraffinwax12345800274211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Paraffin wax

10 Products
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.23
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Agency/Method
  Transition temperature
  Color
  Restriction
  Special Grade
  Melting Point
Show More