• USA Home
  • Search Results

  • paraplast1234514568699311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Paraplast®

1 Products
Synonym:   Paraffin – polyisobutylene mixture
CAS Number:
 MDL number:
 NACRES:
 NA.47
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More