• USA Home
  • Search Results

  • pararosanilinehydrochloride3238256961911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pararosaniline hydrochloride

1 Products
Synonym:   Basic Fuchsin, Basic Parafuchsin, Basic Red 9, Magenta O, Parafuchsin hydrochloride, Paramagenta hydrochloride, Pararosaniline chloride
Image of Pararosaniline hydrochloride
CAS Number:
Linear Formula:
 C19H18N3Cl
Molecular Weight:
 323.82
 Beilstein/REAXYS Number:
 4164603
 Colour Index Number:
 42500
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More