• USA Home
  • Search Results

  • parathyroidhormonefragment134human4117725223267411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Parathyroid Hormone Fragment 1-34 human

1 Products
Synonym:   PTH 1-34, Parathormone (1-34)
CAS Number:
Empirical Formula (Hill Notation):
 C181H291N55O51S2
Molecular Weight:
 4117.72
 MDL number:
 NACRES:
 NA.32
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More