• USA Home
  • Search Results

  • paromomycinsulfatesalt61563freebasebasis126389411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Paromomycin sulfate salt

3 Products
Image of Paromomycin sulfate salt
CAS Number:
Empirical Formula (Hill Notation):
 C23H45N5O14 · xH2SO4
Molecular Weight:
 615.63 (free base basis)
 Beilstein/REAXYS Number:
 5715182
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More