• USA Home
  • Search Results

  • partecksrp80polyvinylalcohol12345900289511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Parteck® SRP 80 Polyvinyl alcohol

1 Products
Synonym:   PVA, PVOH, Parteck® Polyvinyl alcohol
Image of Parteck<SUP>®</SUP> SRP 80 Polyvinyl alcohol
CAS Number:
Linear Formula:
 (C2H4O)n
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
Show More