• USA Home
  • Search Results

  • pbscoretypequantumdots12345131487011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

PbS core-type quantum dots

8 Products
Synonym:   Fluorescent nanocrystals, Lead sulfide, PbS QDs
CAS Number:
Linear Formula:
 PbS
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Fluorescence Test
Show More