• USA Home
  • Search Results

  • pcresol1081410644511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

p-Cresol

5 Products
Synonym:   4-Methylphenol
Image of <I>p</I>-Cresol
CAS Number:
Linear Formula:
 CH3C6H4OH
Molecular Weight:
 108.14
 Beilstein/REAXYS Number:
 1305151
 Council of Europe no.:
 619
 EC Number:
 FEMA Number:
 2337
 Flavis number:
 4.028
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Potency
  Agency/Method
  Melting Point
  Special Grade
Show More