• USA Home
  • Search Results

  • pcymene134229987611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

p-Cymene

5 Products
Synonym:   1-Isopropyl-4-methylbenzene, 4-Isopropyltoluene
Image of <I>p</I>-Cymene
CAS Number:
Linear Formula:
 CH3C6H4CH(CH3)2
Molecular Weight:
 134.22
 Beilstein/REAXYS Number:
 1903377
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Shelf Life
  Physical Form
  Special Grade
  Titertest
Show More