• USA Home
  • Search Results

  • pdoac2322451337531311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

[Pd(OAc)2]3

4 Products
Synonym:   Palladium(II) acetate, Pd(OAc)2
Image of [Pd(OAc)<SUB>2</SUB>]<SUB>3</SUB>
CAS Number:
Linear Formula:
 Pd(OCOCH3)2
Molecular Weight:
 224.51
 Beilstein/REAXYS Number:
 6086766
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Reaction Suitability
  Assay
Show More