• USA Home
  • Search Results

  • pelargoninchloride630981733458611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pelargonin chloride

2 Products
Synonym:   3,5-Bis(glucosyloxy)-4′,7-dihydroxyflavylium chloride, Monardin, Pelargonidin 3,5-di-O-glucoside chloride
Image of Pelargonin chloride
CAS Number:
Empirical Formula (Hill Notation):
 C27H31ClO15
Molecular Weight:
 630.98
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Application(s)
Show More