• USA Home
  • Search Results

  • pemoline17617215234311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pemoline

1 Products
Synonym:   2-Amino-5-phenyl4(5H)-oxazolone, Phenylisohydantoin
Image of Pemoline
CAS Number:
Empirical Formula (Hill Notation):
 C9H8N2O2
Molecular Weight:
 176.17
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
P0048 Pemoline, ≥98% (HPLC),