• USA Home
  • Search Results

  • penicillingpotassiumsalt3724811398411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Penicillin G potassium salt

4 Products
Synonym:   Benzylpenicillin potassium salt
Image of Penicillin G potassium salt
CAS Number:
Empirical Formula (Hill Notation):
 C16H17KN2O4S
Molecular Weight:
 372.48
 Beilstein/REAXYS Number:
 3832841
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 34010400
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Product Line
  Solubility
  Potency
  Application(s)
  Mode of Action
  Physical Form
Show More