• USA Home
  • Search Results

  • penicillingsodiumsalt356376957811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Penicillin G sodium salt

3 Products
Synonym:   Benzylpenicillin sodium salt
Image of Penicillin G sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C16H17N2NaO4S
Molecular Weight:
 356.37
 Beilstein/REAXYS Number:
 3834217
 EC Number:
 MDL number:
 NACRES:
 NA.76
 PubChem Substance ID:
 eCl@ss:
 34010400
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Physical Form
  Solubility
  Mode of Action
Show More