• USA Home
  • Search Results

  • pentaamminechlorocobaltiiichloride250441385951311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentaamminechlorocobalt(III) chloride

1 Products
Synonym:   Cobalt(III) chloropentammine chloride
Image of Pentaamminechlorocobalt(III) chloride
CAS Number:
Linear Formula:
 [Co(NH3)5Cl]Cl2
Molecular Weight:
 250.44
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More