• USA Home
  • Search Results

  • pentacene2783513548811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentacene

4 Products
Image of Pentacene
CAS Number:
Empirical Formula (Hill Notation):
 C22H14
Molecular Weight:
 278.35
 Beilstein/REAXYS Number:
 1912418
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Orbital energy
  Titertest
  OPV Device Performance
  Semiconductor properties
  Solubility
  Special Grade
  Assay
Show More