• USA Home
  • Search Results

  • pentachlorobenzene2503460893511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentachlorobenzene

4 Products
Image of Pentachlorobenzene
CAS Number:
Linear Formula:
 HC6Cl5
Molecular Weight:
 250.34
 Beilstein/REAXYS Number:
 1911550
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Purity Grade
  Anal. Standard Form
  Shelf Life
  Physical Form
  Special Grade
Show More