• USA Home
  • Search Results

  • pentachlorophenol266348786511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentachlorophenol

2 Products
Image of Pentachlorophenol
CAS Number:
Linear Formula:
 C6Cl5OH
Molecular Weight:
 266.34
 Beilstein/REAXYS Number:
 1285380
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Special Grade
  Package quantity
  Physical Form
Show More