• USA Home
  • Search Results

  • pentacontane70334659640311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentacontane

3 Products
Image of Pentacontane
CAS Number:
Linear Formula:
 CH3(CH2)48CH3
Molecular Weight:
 703.34
 Beilstein/REAXYS Number:
 1807879
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
  Special Grade
  Package quantity
Show More