• USA Home
  • Search Results

  • pentadecafluorooctanoicacid4140733567111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentadecafluorooctanoic acid

1 Products
Synonym:   PFOA, Perfluorocaprylic acid, Perfluorooctanoic acid
Image of Pentadecafluorooctanoic acid
CAS Number:
Linear Formula:
 CF3(CF2)6COOH
Molecular Weight:
 414.07
 Beilstein/REAXYS Number:
 1809678
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More