• USA Home
  • Search Results

  • pentadecafluorooctanoicacidsolution4140733567111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentadecafluorooctanoic acid solution

1 Products
Synonym:   PFOA, Perfluorocaprylic acid solution, Perfluorooctanoic acid solution
Image of Pentadecafluorooctanoic acid solution
CAS Number:
Linear Formula:
 CF3(CF2)6COOH
Molecular Weight:
 414.07
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More