• USA Home
  • Search Results

  • pentadecane2124162962911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentadecane

4 Products
Synonym:   n-Pentadecane
Image of Pentadecane
CAS Number:
Linear Formula:
 CH3(CH2)13CH3
Molecular Weight:
 212.41
 Beilstein/REAXYS Number:
 1698194
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Special Grade
  Assay
Show More