• USA Home
  • Search Results

  • pentaerythritol1361511577511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentaerythritol

3 Products
Synonym:   2,2-Bis(hydroxymethyl)-1,3-propanediol
Image of Pentaerythritol
CAS Number:
Linear Formula:
 C(CH2OH)4
Molecular Weight:
 136.15
 Beilstein/REAXYS Number:
 1679274
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
Show More