• USA Home
  • Search Results

  • pentaethyleneglycol23828479215811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentaethylene glycol

1 Products
Synonym:   3,6,9,12-Tetraoxatetradecane-1,14-diol, Pentaglycol
Image of Pentaethylene glycol
CAS Number:
Linear Formula:
 HO(CH2CH2O)5H
Molecular Weight:
 238.28
 Beilstein/REAXYS Number:
 1635593
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Functional group
Show More

Product #
Description
335754 Pentaethylene glycol, 98%,