• USA Home
  • Search Results

  • pentafluorophenol1840677161911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentafluorophenol

2 Products
Synonym:   2,3,4,5,6-Pentafluorophenol
Image of Pentafluorophenol
CAS Number:
Linear Formula:
 C6F5OH
Molecular Weight:
 184.06
 Beilstein/REAXYS Number:
 1912584
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Product Line
  Application(s)
Show More