• USA Home
  • Search Results

  • pentafluoropropionicanhydride3100535642311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentafluoropropionic anhydride

3 Products
Synonym:   PFPA, Perfluoropropionic anhydride
Image of Pentafluoropropionic anhydride
CAS Number:
Linear Formula:
 (CF3CF2CO)2O
Molecular Weight:
 310.05
 Beilstein/REAXYS Number:
 1806446
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Package quantity
  Reaction Suitability
  Assay
Show More