• USA Home
  • Search Results

  • pentane721510966011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentane

11 Products
Image of Pentane
CAS Number:
Linear Formula:
 CH3(CH2)3CH3
Molecular Weight:
 72.15
 Beilstein/REAXYS Number:
 969132
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  Restriction
  Anal. Standard Form
  Transition temperature
  Ice Shipping
  Solubility
  UV-Absorption Condition
  Purity Grade
  Refractive Index
  Application(s)
  Transmittance
  Physical Form
  Special Grade
  Vapor pressure
  Assay
Show More